Learn More
Enterprise Technology Solutions Business Source Laminating Pouch, Letter, 3Mil, 9"x11-1/2", 50/BX, Clear
SDP
Catalog No. 5000740264
Encompass_Preferred
Clear laminating pouch is designed to protect letter-size documents and badges. Short side is sealed. 3 mil pouch offers a standard construction for occasional handling, filing or posting of menus, photos and other items where pouch flexibility is acceptable.
Catalog No. 50-007-40264 Supplier Enterprise Technology Solutions Supplier No. BSN20854
Clear laminating pouch is designed to protect letter-size documents and badges. Short side is sealed. 3 mil pouch offers a standard construction for occasional handling, filing or posting of menus, photos and other items where pouch flexibility is acceptable.