Learn More
Enterprise Technology Solutions Business Source Masking Tape,3" Core,2"x60Yds,1 Roll,Tan
SDP
Catalog No. 5000831812
Encompass_Preferred
Utility-purpose, crepe paper-backed tape is designed for holding, bundling, sealing, noncritical masking and many other jobs where pressure-sensitive tape is needed. Design features a 3" core and approximate tensile strength of 8.5 kg. or 18.7 lb. per "
Catalog No. 50-008-31812 Supplier Enterprise Technology Solutions Supplier No. BSN16442
Utility-purpose, crepe paper-backed tape is designed for holding, bundling, sealing, noncritical masking and many other jobs where pressure-sensitive tape is needed. Design features a 3" core and approximate tensile strength of 8.5 kg. or 18.7 lb. per "