Learn More
Enterprise Technology Solutions Business Source Poly Envelopes, Side Open, Letter,12/PK,Clear
SDP
Catalog No. 5000832371
Encompass_Preferred
Hook-and-loop closure on this poly envelope protects against accidental opening. Envelope expands to hold up to 1" of material. Design is side-opening. Envelope will not transfer print from photocopies.
Catalog No. 50-008-32371 Supplier Enterprise Technology Solutions Supplier No. BSN02017
Hook-and-loop closure on this poly envelope protects against accidental opening. Envelope expands to hold up to 1" of material. Design is side-opening. Envelope will not transfer print from photocopies.