Learn More
Enterprise Technology Solutions Business Source Sheet Protectors,Top Load,3.2 mil,11"x8-1/2",100/BX,Clear
SDP
Catalog No. 5000833203
Encompass_Preferred
Durable, top-loading sheet protectors hold letter-size inserts without punching. Printed materials do not stick or lift off this specially formulated polypropylene material. Design features a reinforced strip at binding edge and three-hole punched sides. Die-cut corners on ring side allow heavyweight pages to turn freely. Oversized ring binders and indexes are sold separately.
Catalog No. 50-008-33203 Supplier Enterprise Technology Solutions Supplier No. BSN74550
Add to Cart
Add to Cart
Durable, top-loading sheet protectors hold letter-size inserts without punching. Printed materials do not stick or lift off this specially formulated polypropylene material. Design features a reinforced strip at binding edge and three-hole punched sides. Die-cut corners on ring side allow heavyweight pages to turn freely. Oversized ring binders and indexes are sold separately.