Learn More
Enterprise Technology Solutions Business Source Stainless Steel Ruler, 12" L, Nonskid, Silver
SDP
Catalog No. 5000825654
Encompass_Preferred
Nonskid ruler is made of stainless steel. Ruler is ruled in 1/16" on one bevel and 1/32" in the first and last " The opposite bevel has metric measurements. All markings and numbers are engraved in black. Ruler has a nonskid cork backing.
Catalog No. 50-008-25654 Supplier Enterprise Technology Solutions Supplier No. BSN32361
Nonskid ruler is made of stainless steel. Ruler is ruled in 1/16" on one bevel and 1/32" in the first and last " The opposite bevel has metric measurements. All markings and numbers are engraved in black. Ruler has a nonskid cork backing.