Learn More
Enterprise Technology Solutions Business Source Storage Box,Heavy-duty,700 lb,Ltr,12"x24"x10",12/CT,WE
SDP
Catalog No. 5000830323
Encompass_Preferred
Heavy-duty storage boxes are ideal for stacking with or without shelving and are designed for maximum stackability. Sturdy, corrugated construction delivers lasting box strength. Average amount of evenly distributed weight that can be safely stacked on top of a box is 700 lb. String-and-button closure keeps letter-size files secure. Storage box contains a high percentage of recycled material.
Catalog No. 50-008-30323 Supplier Enterprise Technology Solutions Supplier No. BSN26756
Heavy-duty storage boxes are ideal for stacking with or without shelving and are designed for maximum stackability. Sturdy, corrugated construction delivers lasting box strength. Average amount of evenly distributed weight that can be safely stacked on top of a box is 700 lb. String-and-button closure keeps letter-size files secure. Storage box contains a high percentage of recycled material.