Learn More
Medchemexpress LLC BUSPIRONE 50.0MG
SDP
Catalog No. NC3806151
Encompass_Preferred
Buspirone is an orally active 5-HT1A receptor agonist, and a dopamine D2 autoreceptorsant antagonist. Buspirone is an anxiolytic agent and can be used for the generalized anxiety disorder research.
Catalog No. NC3806151 Supplier Medchemexpress LLC Supplier No. HYB1115A
Buspirone is an orally active 5-HT1A receptor agonist, and a dopamine D2 autoreceptorsant antagonist. Buspirone is an anxiolytic agent and can be used for the generalized anxiety disorder research.