Learn More
Medchemexpress LLC Buspirone-d8 hydrochloride | 1216761-39-5 | 1 MG
SDP
Catalog No. 5000413071
Encompass_Preferred
Buspirone-d8 (hydrochloride) is the deuterium labeled Buspirone hydrochloride. Buspirone hydrochloride is an anxiolytic psychotropic agent, used to treat generalized anxiety disorder (GAD).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13071 Supplier Medchemexpress LLC Supplier No. HYB1115S1MG
Buspirone-d8 (hydrochloride) is the deuterium labeled Buspirone hydrochloride. Buspirone hydrochloride is an anxiolytic psychotropic agent, used to treat generalized anxiety disorder (GAD).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.