Learn More
Medchemexpress LLC Buspirone-d8 hydrochloride | 1216761-39-5 | CB0105448 | 500 UG
SDP
Catalog No. 5000413319
Encompass_Preferred
Buspirone-d8 hydrochloride is the deuterium labeled Buspirone hydrochloride. It is an anxiolytic psychotropic agent primarily used to treat generalized anxiety disorder (GAD).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13319 Supplier Medchemexpress LLC Supplier No. HYB1115S500UG
Buspirone-d8 hydrochloride is the deuterium labeled Buspirone hydrochloride. It is an anxiolytic psychotropic agent primarily used to treat generalized anxiety disorder (GAD).
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.