Learn More
Apexbio Technology LLC Buspirone HCl 33386-08-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092637
Encompass_Preferred
Buspirone hydrochloride (CAS 33386-08-2) is a selective agonist of the 5-HT1A serotonin receptor subtype engaging central nervous system sites to modulate serotonergic neurotransmission Through its specific interaction with 5-HT1A receptors buspirone hydrochloride influences neurochemical signaling pathways associated with anxiety and mood regulation This compound is widely utilized as a research tool in studies investigating the neurobiological mechanisms underlying anxiety and depression as well as the interplay between central neurotransmitter systems and emotional processing
Catalog No. 50-000-92637 Supplier Apexbio Technology LLC Supplier No. B16875.1
Buspirone hydrochloride (CAS 33386-08-2) is a selective agonist of the 5-HT1A serotonin receptor subtype engaging central nervous system sites to modulate serotonergic neurotransmission Through its specific interaction with 5-HT1A receptors buspirone hydrochloride influences neurochemical signaling pathways associated with anxiety and mood regulation This compound is widely utilized as a research tool in studies investigating the neurobiological mechanisms underlying anxiety and depression as well as the interplay between central neurotransmitter systems and emotional processing