Learn More
Medchemexpress LLC Buspirone (hydrochloride) | 33386-08-2 | 99.99 % | 1 G
SDP
Catalog No. 5000412162
Encompass_Preferred
Buspirone hydrochloride is a 5-HT1A receptor agonist that can be used for anxiety and depression research.
  • Has cytotoxic effect in lymphocytes.
  • Induces ROS formation, MMP collapse, lipid peroxidation, lysosomal damage, and elevation of glutathione disulfide (GSSG) in lymphocytes.
  • Reduces anxiety/depression behaviors in C57BL/6N mice.
  • Restores IS-shifted β-diversity in the gut microbiota in C57BL/6N mice.
  • Reduced TNF-α expression and NF-κB+/Iba1+ cell population in the hippocampus.
  • Reduced myeloperoxidase activity and NF-κB+/CD11c+ cell population in the colon.
  • Reduced the IS- or EC-induced gut proteobacteria population.
Catalog No. 50-004-12162 Supplier Medchemexpress LLC Supplier No. HYB11151G
Buspirone hydrochloride is a 5-HT1A receptor agonist that can be used for anxiety and depression research.
  • Has cytotoxic effect in lymphocytes.
  • Induces ROS formation, MMP collapse, lipid peroxidation, lysosomal damage, and elevation of glutathione disulfide (GSSG) in lymphocytes.
  • Reduces anxiety/depression behaviors in C57BL/6N mice.
  • Restores IS-shifted β-diversity in the gut microbiota in C57BL/6N mice.
  • Reduced TNF-α expression and NF-κB+/Iba1+ cell population in the hippocampus.
  • Reduced myeloperoxidase activity and NF-κB+/CD11c+ cell population in the colon.
  • Reduced the IS- or EC-induced gut proteobacteria population.