Learn More
Medchemexpress LLC Buspirone (hydrochloride) | 33386-08-2 | 99.99% | 250 MG
SDP
Catalog No. 5000414251
Encompass_Preferred
Buspirone hydrochloride is a 5-HT1A receptor agonist used in anxiety and depression research. It has been shown to have cytotoxic effects in lymphocytes and can induce ROS formation, MMP collapse, lipid peroxidation, lysosomal damage, and elevation of glutathione disulfide (GSSG) in these cells. In mice, it has been observed to reduce anxiety/depression behaviors, restore IS-shifted β-diversity in gut microbiota, and reduce inflammation markers in the hippocampus and colon.
  • 5-HT1A receptor agonist
  • Used for anxiety and depression research
  • Can reduce anxiety/depression behaviors in mice
  • Restores IS-shifted β-diversity in the gut microbiota in mice
  • Reduces TNF-α expression and NF-κB+/Iba1+ cell population in the hippocampus and myeloperoxidase activity and NF-κB+/CD11c+ cell population in the colon in mice
Catalog No. 50-004-14251 Supplier Medchemexpress LLC Supplier No. 250MGHYB1115
Buspirone hydrochloride is a 5-HT1A receptor agonist used in anxiety and depression research. It has been shown to have cytotoxic effects in lymphocytes and can induce ROS formation, MMP collapse, lipid peroxidation, lysosomal damage, and elevation of glutathione disulfide (GSSG) in these cells. In mice, it has been observed to reduce anxiety/depression behaviors, restore IS-shifted β-diversity in gut microbiota, and reduce inflammation markers in the hippocampus and colon.
  • 5-HT1A receptor agonist
  • Used for anxiety and depression research
  • Can reduce anxiety/depression behaviors in mice
  • Restores IS-shifted β-diversity in the gut microbiota in mice
  • Reduces TNF-α expression and NF-κB+/Iba1+ cell population in the hippocampus and myeloperoxidase activity and NF-κB+/CD11c+ cell population in the colon in mice