Learn More
Medchemexpress LLC Buspirone hydrochloride | 33386-08-2 | 99.99% | 500 MG
SDP
Catalog No. 5000413300
Encompass_Preferred
Buspirone hydrochloride is a 5-HT1A receptor agonist, commonly utilized in research for anxiety and depression. It demonstrates diverse biological activities, including cytotoxic effects in lymphocytes and modulation of gut microbiota.
  • 5-HT1A receptor agonist.
  • Suitable for anxiety and depression research.
  • Induces cytotoxic effects in lymphocytes.
  • Modulates gut microbiota.
Catalog No. 50-004-13300 Supplier Medchemexpress LLC Supplier No. HYB1115500MG
Buspirone hydrochloride is a 5-HT1A receptor agonist, commonly utilized in research for anxiety and depression. It demonstrates diverse biological activities, including cytotoxic effects in lymphocytes and modulation of gut microbiota.
  • 5-HT1A receptor agonist.
  • Suitable for anxiety and depression research.
  • Induces cytotoxic effects in lymphocytes.
  • Modulates gut microbiota.