Learn More
Cayman Chemical BusRelInNHNH2trIfluoROC 10mg
SDP
Catalog No. 502936547
Encompass_Preferred
A synthetic GnRH agonist; at least 20 times more potent than GNRH; decreases the level of circulating gonadotropins and sex hormones after an initial increase; reduces tumor growth in a DMBA-induced rat model of mammary cancer when used at a dose of 20 UG/kg; induces enteric neurodegeneration, decreasing the number of luteinizing hormone receptor-positive neurons in the rat gastrointestinal tract when administered at a dose of 1 mg/kg for 5 days
Catalog No. 50-293-6547 Supplier Cayman Chemical Supplier No. 2415610 MG
A synthetic GnRH agonist; at least 20 times more potent than GNRH; decreases the level of circulating gonadotropins and sex hormones after an initial increase; reduces tumor growth in a DMBA-induced rat model of mammary cancer when used at a dose of 20 UG/kg; induces enteric neurodegeneration, decreasing the number of luteinizing hormone receptor-positive neurons in the rat gastrointestinal tract when administered at a dose of 1 mg/kg for 5 days