Learn More
STEMCELL Technologies Busulfan, Size: 100 g
SDP
Catalog No. 502390378
Encompass
Inhibit proliferation of cancer cells by creating DNA breaks and DNA crosslinks with busulfan (Iwamoto et al.). Highly potent and cell-permeable busulfan forms interstrand crosslinks between the DNA bases guanine and adenine and between guanine and guanine (Ponti et al.). DNA breakage induced by busulfan-mediated DNA crosslinking induces senescence in cells (Chen et al.).
Catalog No. 50-239-0378 Supplier STEMCELL Technologies Supplier No. 1001125
Inhibit proliferation of cancer cells by creating DNA breaks and DNA crosslinks with busulfan (Iwamoto et al.). Highly potent and cell-permeable busulfan forms interstrand crosslinks between the DNA bases guanine and adenine and between guanine and guanine (Ponti et al.). DNA breakage induced by busulfan-mediated DNA crosslinking induces senescence in cells (Chen et al.).