Learn More
Apexbio Technology LLC Busulfan, 1g. Cas: 55-98-1 MFCD: MFCD00007562. DNA alkylating agent
SDP
Supplier:  Apexbio Technology LLC A83861000
Encompass_Preferred
Antineoplastic bifunctional, DNA alkylating agent that causes DNA damage by cross-linking DNA and proteins. Causes double-base lesions mainly at 5'-GA-3' and, to a lesser extent, at 5'-GG-3' sequences. 55-98-1. MFCD00007562 Busulfan is a DNA alkylating agent [1]. DNA alkylating agent is attached to the guanine base of DNA and stops tumor growth by crosslinking guanine nucleobases in DNA double-helix strands and causes DNA damage [1]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8593