Learn More
Medchemexpress LLC Busulfan | 55-98-1 | 99.78% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB024510G
Encompass_Preferred
Busulfan is a potent alkylating antineoplastic agent. It causes DNA damage by cross-linking DNAs and both DNA and proteins. Additionally, Busulfan inhibits thioredoxin reductase and is known to induce apoptosis. This compound functions as an immunosuppressive and myeloablative chemotherapeutic agent.
  • Potent alkylating antineoplastic agent
  • Causes DNA damage by cross-linking DNAs and DNA and proteins
  • Inhibits thioredoxin reductase
  • Induces apoptosis
  • Immunosuppressive chemotherapeutic agent
  • Myeloablative chemotherapeutic agent
Catalog No. 50-004-05617