Learn More
Medchemexpress LLC Busulfan-d8 | 116653-28-2 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0245S1MG
Encompass_Preferred
Busulfan-d8 is a deuterium labeled Busulfan, functioning as a potent alkylating antineoplastic agent. It causes DNA damage by cross-linking DNAs and DNA and proteins, inhibits thioredoxin reductase, and induces apoptosis. This compound acts as an immunosuppressive and myeloablative chemotherapeutic agent.
  • Deuterium labeled compound
  • Functions as a potent alkylating antineoplastic agent
  • Causes DNA damage by cross-linking DNAs and DNA and proteins
  • Inhibits thioredoxin reductase and induces apoptosis
  • Acts as an immunosuppressive and myeloablative chemotherapeutic agent
  • Can be used as a tracer for quantitation during drug development
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06183