Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Busulfan-d8 | 116653-28-2 | MFCD06658695 | 99.7% | 254.35 | C6H6D8O6S2 | 10 MG
SDP

Catalog No. 5000297072
Encompass_Preferred

Deuterium-labeled busulfan for use as an internal standard and tracer in quantitative analytical methods. It is supplied as a solid with high chemical and isotopic purity and is intended for research use and therapeutic drug monitoring.

  • Intended for use as an internal standard and tracer in NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular formula C6H6D8O6S2 and molecular weight 254.35.
  • High purity (≈99.7%) and white to off-white solid appearance.
  • Available in a 10 mg quantity suitable for analytical applications.
  • Storage recommendations: powder at -20°C for long-term stability; in solvent at -80°C for short-term storage.

Catalog No. 50-002-97072 Supplier Medchemexpress LLC Supplier No. HYB0245S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled busulfan for use as an internal standard and tracer in quantitative analytical methods. It is supplied as a solid with high chemical and isotopic purity and is intended for research use and therapeutic drug monitoring.

  • Intended for use as an internal standard and tracer in NMR, GC-MS, and LC-MS quantitative analysis.
  • Molecular formula C6H6D8O6S2 and molecular weight 254.35.
  • High purity (≈99.7%) and white to off-white solid appearance.
  • Available in a 10 mg quantity suitable for analytical applications.
  • Storage recommendations: powder at -20°C for long-term stability; in solvent at -80°C for short-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.