Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Busulfan, MilliporeSigma™ Supelco™

Catalog No. 111017872 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7872 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7872 Supplier MilliporeSigma Supelco Supplier No. B263525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Busulfan is a bifunctional alkylating agent, which may find application in the treatment of chronic granulocytic leukemia.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 55-98-1
Physical Form Neat
Linear Formula CH3 SO2 O(CH2)4 OSO2 CH3
MDL Number MFCD00007562
Melting Point 114°C to 117°C
Synonym 1,4-Butanediol dimethanesulfonate
Type Analytical Standard
Industry Type Drug Analysis
Quantity 25 g
Formula Weight 246.30
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.