Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem But-2-enoic acid | ≥95% | CAS 107-93-7 | C4H6O2 | 1kg

Catalog No. 5001039044
Encompass_Preferred

But-2-enoic acid | ≥95% | CAS 107-93-7 | C4H6O2 | 1kg

Catalog No. 50-010-39044 Supplier Aobchem Supplier No. A5188981KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

But-2-enoic acid | ≥95% | CAS 107-93-7 | C4H6O2 | 1kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.