Learn More
Medchemexpress LLC Butabindide oxalate | 185213-03-0 | MFCD09971008 | 99.0% | 393.43 g/mol | C19H27N3O6 | 5 MG
SDP
Catalog No. 5000241197
Encompass_Preferred
Butabindide oxalate is a selective, potent inhibitor of tripeptidyl peptidase II (TPP II) used in biochemical and pharmacological research to inhibit TPP II activity and protect neuropeptides such as cholecystokinin-8 (CCK-8) from enzymatic degradation.
  • Potent TPP II inhibition with Ki ≈ 7 nM.
  • High selectivity over TPP I (Ki ≈ 10 μM).
  • Supplied as an oxalate salt, white to off-white solid.
  • High purity suitable for biochemical assays (≈99%).
  • Available in small, laboratory-scale vials for research use.
Catalog No. 50-002-41197 Supplier Medchemexpress LLC Supplier No. HY1077595MG
Butabindide oxalate is a selective, potent inhibitor of tripeptidyl peptidase II (TPP II) used in biochemical and pharmacological research to inhibit TPP II activity and protect neuropeptides such as cholecystokinin-8 (CCK-8) from enzymatic degradation.
  • Potent TPP II inhibition with Ki ≈ 7 nM.
  • High selectivity over TPP I (Ki ≈ 10 μM).
  • Supplied as an oxalate salt, white to off-white solid.
  • High purity suitable for biochemical assays (≈99%).
  • Available in small, laboratory-scale vials for research use.