Learn More
Medchemexpress LLC Butabindide oxalate 1mg | 185213-03-0 | 1MG
SDP
Catalog No. 5000240325
Encompass_Preferred
Butabindide (UCL-1397) oxalate is a potent selective tripeptidvl peptidase II (TPP II) inhibitor with Ki values of 7 nM and 10 M for TPP II and TPP I respectively Butabindide oxalate inhibits TPP II to protect CCK-8 against inactivation[1 [2
Catalog No. 50-002-40325 Supplier Medchemexpress LLC Supplier No. HY1077591MG
Butabindide (UCL-1397) oxalate is a potent selective tripeptidvl peptidase II (TPP II) inhibitor with Ki values of 7 nM and 10 M for TPP II and TPP I respectively Butabindide oxalate inhibits TPP II to protect CCK-8 against inactivation[1 [2