Learn More
Cayman Chemical Butabindide (oxalate), Purity: ≥98%, CAS: 185213-03-0, 10 mg
SDP
Catalog No. 502987470
Encompass_Preferred
A potent inhibitor of CCK-inactivating peptidase/TPP-2 (Ki = 7 nM); selective for CCK-inactivating peptidase/TPP-2 over a panel of serine proteases (Kis = >1 μM) as well as CCK receptors (Kis = >0.1 mM); increases levels of CCK octapeptide in depolarized rat cerebral cortex slices; inhibits CCK-inactivating peptidase/TPP-2 in mouse liver and brain (ID50s = 1.1 and 6.8 mg/kg, respectively); enhances CCK octapeptide-induced delay in gastric emptying and reduces food intake in mice
Catalog No. 50-298-7470 Supplier Cayman Chemical Supplier No. 2161010 MG
A potent inhibitor of CCK-inactivating peptidase/TPP-2 (Ki = 7 nM); selective for CCK-inactivating peptidase/TPP-2 over a panel of serine proteases (Kis = >1 μM) as well as CCK receptors (Kis = >0.1 mM); increases levels of CCK octapeptide in depolarized rat cerebral cortex slices; inhibits CCK-inactivating peptidase/TPP-2 in mouse liver and brain (ID50s = 1.1 and 6.8 mg/kg, respectively); enhances CCK octapeptide-induced delay in gastric emptying and reduces food intake in mice