Learn More
Butadiene diepoxide, 95%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Butadiene diepoxide is used as a research chemical, as a chemical intermediate, as a curing agent for polymers, as a cross-linking agent for textile fabrics, and to prevent microbial spoilage. It is also been used to synthesize erythritol and pharmaceuticals.
Solubility
Soluble in water (1000 g/L).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -19°C |
| Density | 1.112 |
| Boiling Point | 56°C to 58°C (25 mmHg) |
| Flash Point | 45°C (113°F) |
| Refractive Index | 1.434 |
| Quantity | 1 g |
| UN Number | UN2929 |
| Beilstein | 79831 |
| Merck Index | 14,3676 |
| Solubility Information | Soluble in water (1000g/L). |
| Show More |
Safety and Handling
| Hazard Category | H226-H301-H310+H330-H314-H335-H340-H350-H373 |
| Hazard Statement | GHS H Statement H301-H311-H330-H340-H350-H314-H226 Toxic if swallowed. Toxic in contact with skin. Fatal if inhaled. May cause genetic defects. May cause cancer. Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: I |
| EINECSNumber | 215-979-1 |
| RTECSNumber | EJ8400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.