Learn More
Medchemexpress LLC Butamben | 94-25-7 | 99.8% | 10 G
SDP
Catalog No. 5000412912
Encompass_Preferred
Butamben (Butyl 4-aminobenzoate) provides long-lasting pain relief without impairing motor function or other sensory functions. This compound is used in research for its effects on ion channels.
  • Blocks barium current in PC12 cells
  • Increases inactivation of fast Na+ channels
  • Exhibits analgesic activity in mice tail-flick assay
Catalog No. 50-004-12912 Supplier Medchemexpress LLC Supplier No. HYB143010G
Butamben (Butyl 4-aminobenzoate) provides long-lasting pain relief without impairing motor function or other sensory functions. This compound is used in research for its effects on ion channels.
  • Blocks barium current in PC12 cells
  • Increases inactivation of fast Na+ channels
  • Exhibits analgesic activity in mice tail-flick assay