Learn More
Medchemexpress LLC Butamben (butyl 4-aminobenzoate) | 94-25-7 | 99.82% | 1 ML
SDP
Catalog No. 5000413506
Encompass_Preferred
Butamben (Butyl 4-aminobenzoate) is a calcium channel inhibitor that provides long-lasting relief from pain without impairing motor or other sensory functions. It is intended for research use only.
  • Blocks 90% of control barium current in PC12 cells
  • Increases inactivation of fast Na+ channels
  • Shows dose-dependent analgesic activity in mice
Catalog No. 50-004-13506 Supplier Medchemexpress LLC Supplier No. HYB143010MM/1ML
Butamben (Butyl 4-aminobenzoate) is a calcium channel inhibitor that provides long-lasting relief from pain without impairing motor or other sensory functions. It is intended for research use only.
  • Blocks 90% of control barium current in PC12 cells
  • Increases inactivation of fast Na+ channels
  • Shows dose-dependent analgesic activity in mice