Learn More
Medchemexpress LLC Butanamide, N-(2,6-dimethylphenyl)-2-(ethylpropylamino)-, hydrochloride (1:1) | 36637-19-1 | 99.9% | 25 MG
SDP
Catalog No. 5000411636
Encompass_Preferred
Etidocaine hydrochloride is a long aminoamide local agent designed to suppress or relieve pain, intended for research use only.
Key Features:
  • Functions as a long aminoamide local agent
  • Effective in pain suppression or relief
  • Provided as a white to off-white solid
  • High purity of 99.93%
  • Soluble in DMSO at 62.5 mg/mL
  • Stable as a powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month
Catalog No. 50-004-11636 Supplier Medchemexpress LLC Supplier No. 25MGHYB2080A
Add to Cart
Add to Cart
Etidocaine hydrochloride is a long aminoamide local agent designed to suppress or relieve pain, intended for research use only.
Key Features:
  • Functions as a long aminoamide local agent
  • Effective in pain suppression or relief
  • Provided as a white to off-white solid
  • High purity of 99.93%
  • Soluble in DMSO at 62.5 mg/mL
  • Stable as a powder at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month