Learn More
U.S. Pharmacopeia Butane-1,3-diol, 107-88-0, MFCD00004554, 1mL
Catalog No. NC1358671
Encompass
Molecular formula C4H10O2, Molecular weight 90.12, Melting Point < 122 °F (< 50 °C), Boiling Point 405.5 °F (207.5 °C) 101.325 kPa, Synonyms: 1,3-Butandiol, Butylene glycol
Catalog No. NC1358671 Supplier U.S. Pharmacopeia Supplier No. 1082956
Molecular formula C4H10O2, Molecular weight 90.12, Melting Point < 122 °F (< 50 °C), Boiling Point 405.5 °F (207.5 °C) 101.325 kPa, Synonyms: 1,3-Butandiol, Butylene glycol