Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Butane-1 4-diammonium Iodide

Catalog No. 91282410G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-282-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-282-410G Supplier Sigma Aldrich Supplier No. 91282410G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Organohalide based perovskites have emerged as an important class of material for solar cell applications. The variations/substitution in organohalide cations and anions is employed for the optimization of the band gap, carrier diffusion length, and power conversion efficiency of perovskites based solar cells. Therefore adding a suite of organohalide precursors would be a good expansion of our Perovskites portfolio.Product of Greatcell Solar(R)

Specifications

CAS 916849-52-0
Molecular Formula C4H14I2N2
Linear Formula C4H14I2N2
Quantity 10 g
MDL Number MFCD32062897
Synonym 1,4-Butanediamine dihydriodide; 1,4-Butanediamine dihydroiodide; 1,4-Diaminobutane dihydriodide; Butane-1,4-diaminium iodide; Butane-1,4-diamino dihydriodide; NBUDAI
Molecular Weight (g/mol) 343.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.