Learn More
Apexbio Technology LLC Butane diacid 110-15-6 100mg
SDP
Catalog No. 5000090096
Encompass_Preferred
Butane diacid also known as succinic acid (CAS 110-15-6) is a saturated dicarboxylic acid commonly utilized in biomedical research It participates in cellular metabolic pathways including the tricarboxylic acid (TCA) cycle where it functions as an intermediate involved in energy production and metabolic regulation Butane diacid is frequently employed to study mitochondrial respiration cellular energy metabolism and to investigate metabolic disorders The compound is water-soluble and should be stored in tightly sealed containers under cool dry conditions to maintain stability during experimental use
Catalog No. 50-000-90096 Supplier Apexbio Technology LLC Supplier No. N1941100
Butane diacid also known as succinic acid (CAS 110-15-6) is a saturated dicarboxylic acid commonly utilized in biomedical research It participates in cellular metabolic pathways including the tricarboxylic acid (TCA) cycle where it functions as an intermediate involved in energy production and metabolic regulation Butane diacid is frequently employed to study mitochondrial respiration cellular energy metabolism and to investigate metabolic disorders The compound is water-soluble and should be stored in tightly sealed containers under cool dry conditions to maintain stability during experimental use