Learn More
Medchemexpress LLC Butanedioic-1,2,3,4-13C4 acid | 201595-67-7 | 122.06 | 5 MG
SDP
Catalog No. 5000389655
Encompass_Preferred
Succinic acid-13C4 is the 13C labeled succinic acid. It acts as a potent anxiolytic agent and is an intermediate product of the tricarboxylic acid cycle. This compound serves as a precursor for various industrially important chemicals across food, chemical, and pharmaceutical sectors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89655 Supplier Medchemexpress LLC Supplier No. HYN0420S25MG
Succinic acid-13C4 is the 13C labeled succinic acid. It acts as a potent anxiolytic agent and is an intermediate product of the tricarboxylic acid cycle. This compound serves as a precursor for various industrially important chemicals across food, chemical, and pharmaceutical sectors.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.