Learn More
Medchemexpress LLC Butanedioic acid-1,4-13C2 | 79864-95-2 | MFCD00224213 | 99.0% | 120.07 | C4H6O4 | 10 MG
SDP
Catalog No. 5000295816
Encompass_Preferred
Succinic acid-1,4-13C2 is a stable isotope-labeled form of succinic acid supplied as a white to off-white solid for research and analytical use, including metabolic tracing and mass spectrometry standards. The material is high purity and intended for analytical applications.
  • Stable isotope-labeled succinic acid (1,4-13C2) for tracer studies and internal standards.
  • High purity (99.0%) suitable for analytical and mass spectrometry applications.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Suitable for metabolic tracing, quantitation, and method validation.
  • Recommended storage: powder at -20°C for long-term stability; short-term storage at 4°C.
Catalog No. 50-002-95816 Supplier Medchemexpress LLC Supplier No. HYN0420S310MG
Succinic acid-1,4-13C2 is a stable isotope-labeled form of succinic acid supplied as a white to off-white solid for research and analytical use, including metabolic tracing and mass spectrometry standards. The material is high purity and intended for analytical applications.
  • Stable isotope-labeled succinic acid (1,4-13C2) for tracer studies and internal standards.
  • High purity (99.0%) suitable for analytical and mass spectrometry applications.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Suitable for metabolic tracing, quantitation, and method validation.
  • Recommended storage: powder at -20°C for long-term stability; short-term storage at 4°C.