Learn More
Medchemexpress LLC Butanoic-1-13C acid, 3-methyl- | 87994-84-1 | 99.0% | 103.12 | 5 MG
SDP
Catalog No. 5000366753
Encompass_Preferred
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and is known to affect neonatal death and possible Jamaican vomiting sickness in humans. This product is for research use only and is not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-66753 Supplier Medchemexpress LLC Supplier No. HYW012980S15MG
Add to Cart
Add to Cart
Isovaleric acid-13C is the 13C labeled Isovaleric acid. Isovaleric acid is a natural fatty acid and is known to affect neonatal death and possible Jamaican vomiting sickness in humans. This product is for research use only and is not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS