Learn More
Medchemexpress LLC Butanoic-2,2,4,4,4-d5 acid, 3-oxo-, ethyl ester | 55514-60-8 | 99.2% | 135.17 | C6H5D5O3 | 25mg
SDP
Catalog No. 5000267490
Encompass_Preferred
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]
Catalog No. 50-002-67490 Supplier Medchemexpress LLC Supplier No. HYY1093S225MG
Ethyl acetoacetate-d5 is the deuterium labeled Ethyl acetoacetate[1] Ethyl acetoacetate (Ethyl acetylacetate) is an ester widely used as an intermediate in the synthesis of many varieties of compounds[2][3][4] Ethyl acetoacetate is an inhibitor of bacterial biofilm[5]