Learn More
Medchemexpress LLC Butanoic acid, 2-amino-4-azido-, hydrochloride (1:1), (2R)- | 1858224-26-6 | C4H9ClN4O2 | 25 MG
SDP
Catalog No. 5000374532
Encompass_Preferred
H-D-Aha-OH hydrochloride is a click chemistry reagent that contains an azide group. It is designed for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and for ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. This product is for research use only.
  • Contains an azide group
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Functions as an ADC linker
  • Suitable for antibody-drug conjugate related research
  • For research use only
Catalog No. 50-003-74532 Supplier Medchemexpress LLC Supplier No. 25MGHY151723
H-D-Aha-OH hydrochloride is a click chemistry reagent that contains an azide group. It is designed for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and for ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. This product is for research use only.
  • Contains an azide group
  • Compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Compatible with ring strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Functions as an ADC linker
  • Suitable for antibody-drug conjugate related research
  • For research use only