Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butanoic acid, 2-ethyl-2-hydroxy- | 3639-21-2 | 98.0% | C6H12O3 | 1 G
SDP

Catalog No. 5000353693
Encompass_Preferred

2-Ethyl-2-hydroxybutanoic acid, also known as 2-Ethyl-2-hydroxybutyric acid, is a hydroxy fatty acid. It is identified as a metabolite of DEHP and is provided as a white to off-white solid.

  • Hydroxy fatty acid
  • Metabolite of DEHP
  • High purity (98.0%)
  • Consistent with structure
  • Suitable for research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-53693 Supplier Medchemexpress LLC Supplier No. HY1421051G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Ethyl-2-hydroxybutanoic acid, also known as 2-Ethyl-2-hydroxybutyric acid, is a hydroxy fatty acid. It is identified as a metabolite of DEHP and is provided as a white to off-white solid.

  • Hydroxy fatty acid
  • Metabolite of DEHP
  • High purity (98.0%)
  • Consistent with structure
  • Suitable for research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.