Learn More
Medchemexpress LLC Butanoic acid, 2-hydroxy-, (2R)- | 20016-85-7 | 99.9% | 104.10 | 250 MG
SDP
Catalog No. 5000439900
Encompass_Preferred
(R)-2-Hydroxybutanoic acid is an isomer of 2-Hydroxybutyric acid and can serve as an experimental control. This compound is a byproduct of insulin resistance, which can lead to impaired insulin signaling and mitochondrial overload. It enhances the viability of BRL3A, AML12, and primary mouse hepatocytes, and reduces AP-induced cytotoxicity.
  • Isomer of 2-hydroxybutyric acid
  • Can be used as an experimental control
  • Enhances cell viability and reduces cytotoxicity in certain cell lines
  • Appearance: solid (<50°C), liquid (>54°C)
  • Color: colorless to off-white
Catalog No. 50-004-39900 Supplier Medchemexpress LLC Supplier No. 250MGHY113381A
(R)-2-Hydroxybutanoic acid is an isomer of 2-Hydroxybutyric acid and can serve as an experimental control. This compound is a byproduct of insulin resistance, which can lead to impaired insulin signaling and mitochondrial overload. It enhances the viability of BRL3A, AML12, and primary mouse hepatocytes, and reduces AP-induced cytotoxicity.
  • Isomer of 2-hydroxybutyric acid
  • Can be used as an experimental control
  • Enhances cell viability and reduces cytotoxicity in certain cell lines
  • Appearance: solid (<50°C), liquid (>54°C)
  • Color: colorless to off-white