Learn More
Medchemexpress LLC Butanoic acid,3-hydroxy- | 300-85-6 | 99.8% | 104.10 | 1 G
SDP
Catalog No. 5000436634
Encompass_Preferred
3-Hydroxybutyric acid is an endogenous metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids, interacting with lipid monolayers and altering phase behavior at clinical concentrations.
  • Endogenous metabolite
  • Interacts with lipid monolayers and alters phase behavior at clinical concentrations
  • Diminishes the interfacial viscosity of DPPC Monolayers
  • Appearance: <33°C Solid, >34°C Liquid
  • Color: Off-white to light yellow
  • Initial source: Microorganisms
Catalog No. 50-004-36634 Supplier Medchemexpress LLC Supplier No. HY1133781G
3-Hydroxybutyric acid is an endogenous metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids, interacting with lipid monolayers and altering phase behavior at clinical concentrations.
  • Endogenous metabolite
  • Interacts with lipid monolayers and alters phase behavior at clinical concentrations
  • Diminishes the interfacial viscosity of DPPC Monolayers
  • Appearance: <33°C Solid, >34°C Liquid
  • Color: Off-white to light yellow
  • Initial source: Microorganisms