Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butanoic acid, 3-methyl-, 5-methyl-2-(1-methylethyl)cyclohexyl ester | 16409-46-4 | MFCD00045488 | 98.0% | 240.38 g/mol | C15H28O2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW68386410MM/1ML

Encompass_Preferred

Menthyl isovalerate is an ester of menthol and isovaleric acid supplied as a ready-to-use solution for research and flavor/fragrance applications. The product is formulated at a 10 mM concentration in dimethyl sulfoxide (DMSO) and is provided in a single 1 mL vial. Analytical documentation including a datasheet, certificate of analysis, and safety data sheet are available for quality and handling information.

  • Supplied as a 10 mM solution in dimethyl sulfoxide (DMSO) in a 1 mL vial.
  • Purity 98.0% by NMR, with certificate of analysis available.
  • Molecular formula C15H28O2 and molecular weight 240.38 g/mol.
  • CAS number 16409-46-4 for unambiguous chemical identification.
  • Datasheet, COA, and SDS available to support safe handling and quality assurance.

Catalog No. 50-002-84045


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.