Learn More
Medchemexpress LLC Butanoic acid, 4-[[(1,1-dimethylethoxy)carbonyl]amino]-2-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-, (2S)- | 125238-99-5 | 440.49 | 1 ML
SDP
Catalog No. 5000447965
Encompass_Preferred
Fmoc-Dab(Boc)-OH is an amino acid derivative with an Fmoc protecting group. It can be used to synthesize somatostatin analogs that inhibit neointima formation induced by balloon injury in rats without altering growth hormone release, and can also be used for the synthesis of peptides.
  • Amino acid derivative with an Fmoc protecting group
  • Synthesizes somatostatin analogs
  • Inhibits neointima formation induced by balloon injury in rats
  • Does not alter growth hormone release
  • Used for peptide synthesis
Catalog No. 50-004-47965 Supplier Medchemexpress LLC Supplier No. HYW00802410MM/1ML
Fmoc-Dab(Boc)-OH is an amino acid derivative with an Fmoc protecting group. It can be used to synthesize somatostatin analogs that inhibit neointima formation induced by balloon injury in rats without altering growth hormone release, and can also be used for the synthesis of peptides.
  • Amino acid derivative with an Fmoc protecting group
  • Synthesizes somatostatin analogs
  • Inhibits neointima formation induced by balloon injury in rats
  • Does not alter growth hormone release
  • Used for peptide synthesis