Learn More
Medchemexpress LLC Butanoic acid, 4-[[2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetyl | 2308035-51-8 | 98.4% | 417.37 g/mol | C19H19N3O8 | 1 G
SDP
Catalog No. 5000244795
Encompass_Preferred
Thalidomide-O-amido-C3-COOH is an E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a short carboxylic-acid linker for use in PROTAC design, synthesis, and degradation studies.
  • Used as a cereblon E3 ligase ligand in PROTAC design and synthesis.
  • High purity (98.4%) suitable for research applications.
  • White to off-white solid with molecular weight 417.37 g/mol.
  • Contains a carboxylic acid functional handle for conjugation chemistry.
  • Available in small lab-scale quantities and supplied with a certificate of analysis.
Catalog No. 50-002-44795 Supplier Medchemexpress LLC Supplier No. HY1036121G
Thalidomide-O-amido-C3-COOH is an E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon ligand with a short carboxylic-acid linker for use in PROTAC design, synthesis, and degradation studies.
  • Used as a cereblon E3 ligase ligand in PROTAC design and synthesis.
  • High purity (98.4%) suitable for research applications.
  • White to off-white solid with molecular weight 417.37 g/mol.
  • Contains a carboxylic acid functional handle for conjugation chemistry.
  • Available in small lab-scale quantities and supplied with a certificate of analysis.