Learn More
Medchemexpress LLC Butanoic acid, 4-[3-[[1-(4-chlorophenyl)-2-[2,3-dihydro-6-(trifluoromethoxy)-1H-indol-1-yl]-2-oxoethyl]amino]-5-methoxyphenoxy]-, (-)- | 2135640-92-3 | 99.35% | 578.96 | 1 MG
SDP
Catalog No. 5000348267
Encompass_Preferred
(-)-JNJ-A07 is a potent and selective DENV inhibitor with an EC50 value of 31 nM. It is for research use only and not sold to patients.
  • For research use only
  • Potent and selective DENV inhibitor with an EC50 value of 31 nM
  • Appearance: white to light yellow solid
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-48267 Supplier Medchemexpress LLC Supplier No. HY139602C1MG
(-)-JNJ-A07 is a potent and selective DENV inhibitor with an EC50 value of 31 nM. It is for research use only and not sold to patients.
  • For research use only
  • Potent and selective DENV inhibitor with an EC50 value of 31 nM
  • Appearance: white to light yellow solid
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month