Learn More
Medchemexpress LLC Butanoic acid, 4-[3-[[1-(4-chlorophenyl)-2-[2,3-dihydro-6-(trifluoromethoxy)-1H-indol-1-yl]... | 2135640-92-3 | 98.0% | C28H26ClF3N2O6 | 10MG
SDP
Catalog No. 5000259893
Encompass_Preferred
(-)-JNJ-A07 is an enantiomeric small-molecule inhibitor of dengue virus (DENV) replication, reported to have an EC50 of 31 nM. Supplied as a purified solid (CAS 2135640-92-3) with molecular formula C28H26ClF3N2O6, it is suitable for in vitro and in vivo research and can be formulated in DMSO or combination vehicles for dosing.
Catalog No. 50-002-59893 Supplier Medchemexpress LLC Supplier No. HY139602C10MG
(-)-JNJ-A07 is an enantiomeric small-molecule inhibitor of dengue virus (DENV) replication, reported to have an EC50 of 31 nM. Supplied as a purified solid (CAS 2135640-92-3) with molecular formula C28H26ClF3N2O6, it is suitable for in vitro and in vivo research and can be formulated in DMSO or combination vehicles for dosing.