Learn More
Medchemexpress LLC Butanoic acid, 4-amino-3-[[(9H-fluoren-9-ylmethoxy)carbonyl]amino]-4-oxo-, (3S)- | 200335-40-6 | MFCD00151920 | 98.5% | 354.36 | C19H18N2O5 | 100 MG
SDP
Catalog No. 5000202430
Encompass_Preferred
Fmoc-Asp-NH2 is a cleavable antibody-drug conjugate (ADC) linker consisting of an Fmoc-protected aspartic acid amide. Supplied as a white to off-white solid for research use only, it is used in linker assembly and conjugation steps during ADC and peptide synthesis.
  • Cleavable linker suitable for ADC synthesis.
  • Fmoc-protected aspartamide functionality for peptide and linker assembly.
  • High purity (typical 98.5%); individual batch COA may vary.
  • DMSO solubility: 250 mg/mL; ultrasonic treatment recommended for dissolution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-02430 Supplier Medchemexpress LLC Supplier No. HY135418100MG
Fmoc-Asp-NH2 is a cleavable antibody-drug conjugate (ADC) linker consisting of an Fmoc-protected aspartic acid amide. Supplied as a white to off-white solid for research use only, it is used in linker assembly and conjugation steps during ADC and peptide synthesis.
  • Cleavable linker suitable for ADC synthesis.
  • Fmoc-protected aspartamide functionality for peptide and linker assembly.
  • High purity (typical 98.5%); individual batch COA may vary.
  • DMSO solubility: 250 mg/mL; ultrasonic treatment recommended for dissolution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).