Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butanoic acid, 4-azido- | 54447-68-6 | MFCD11850695 | 99.5% | 129.12 g/mol | C4H7N3O2 | 500 MG
SDP

Catalog No. 5000284804
Encompass_Preferred

4-Azidobutyric acid is an azide-containing carboxylic acid used as an intermediate in organic synthesis and chemical biology research. Supplied as a 500 mg reagent, analytical data are provided to confirm identity and purity.

  • High purity (99.48% GC).
  • Molecular formula C4H7N3O2; molecular weight 129.12 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Packaged as a 500 mg vial for laboratory use.
  • Storage: -20°C for up to 3 years (pure form); in solvent store at -80°C for long-term.
  • Analytical data provided: 1H NMR and LCMS consistent with structure.

Catalog No. 50-002-84804 Supplier Medchemexpress LLC Supplier No. HYW588704500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Azidobutyric acid is an azide-containing carboxylic acid used as an intermediate in organic synthesis and chemical biology research. Supplied as a 500 mg reagent, analytical data are provided to confirm identity and purity.

  • High purity (99.48% GC).
  • Molecular formula C4H7N3O2; molecular weight 129.12 g/mol.
  • Appearance: colorless to light yellow liquid.
  • Packaged as a 500 mg vial for laboratory use.
  • Storage: -20°C for up to 3 years (pure form); in solvent store at -80°C for long-term.
  • Analytical data provided: 1H NMR and LCMS consistent with structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.