Learn More
Medchemexpress LLC Butanoic acid NHS ester (BHQ-1) | 916753-61-2 | 95.4% | 601.61 g/mol | C30H31N7O7 | 10 MG
SDP
Catalog No. 5000287433
Encompass_Preferred
BHQ-1 NHS is an N-hydroxysuccinimide (NHS) ester derivative of a non-fluorescent quencher designed for covalent coupling to primary amines on nucleotides, oligonucleotides, and other biomolecules. It is intended for use as a quencher in fluorescence probe and assay design and for direct labeling of fluorescently labeled nucleotides. For research use only.
  • Reactive NHS ester for stable amide bond formation with amines.
  • Non-fluorescent quenching suitable for fluorescence-based assays.
  • High purity, typically 95.44% as stated on the COA.
  • Soluble in DMSO at approximately 4.35 mg/mL; ultrasonic agitation recommended.
  • Provided in milligram quantities for small-scale labeling workflows.
Catalog No. 50-002-87433 Supplier Medchemexpress LLC Supplier No. HY15483410MG
BHQ-1 NHS is an N-hydroxysuccinimide (NHS) ester derivative of a non-fluorescent quencher designed for covalent coupling to primary amines on nucleotides, oligonucleotides, and other biomolecules. It is intended for use as a quencher in fluorescence probe and assay design and for direct labeling of fluorescently labeled nucleotides. For research use only.
  • Reactive NHS ester for stable amide bond formation with amines.
  • Non-fluorescent quenching suitable for fluorescence-based assays.
  • High purity, typically 95.44% as stated on the COA.
  • Soluble in DMSO at approximately 4.35 mg/mL; ultrasonic agitation recommended.
  • Provided in milligram quantities for small-scale labeling workflows.