Learn More
Crescent Chemical Co Inc BUTANOL MOLECULAR BIOLOGY GRDE
SDP
Catalog No. 50248376
Encompass_Preferred
Butanol molecular biology grade n-Butanol is used for the removal of ethidium bromide from DNA purified by CsCl gradient ultracentrifugation. It is also useful for the concentration of diluted nucleic acid solutions by repeated extractions. Size - 500
Catalog No. 50-248-376 Supplier Crescent Chemical Co Inc Supplier No. SE39550.01
Butanol molecular biology grade n-Butanol is used for the removal of ethidium bromide from DNA purified by CsCl gradient ultracentrifugation. It is also useful for the concentration of diluted nucleic acid solutions by repeated extractions. Size - 500