Learn More
Cayman Chemical Butaprost (free acid), Purity: ≥95%, CAS: 433219-55-7, 10 mg
SDP
Catalog No. 502953352
Encompass_Preferred
A potent, selective EP2 receptor agonist (Ki = 73 nM)
Catalog No. 50-295-3352 Supplier Cayman Chemical Supplier No. 1374110 MG
A potent, selective EP2 receptor agonist (Ki = 73 nM)