Learn More
Medchemexpress LLC Butaxamine hydrochloride | 5696-15-1 | 99.9% | 303.82 | C15H26ClNO3 | 10 MG
SDP
Catalog No. 5000227456
Encompass_Preferred
Butaxamine hydrochloride is the hydrochloride salt of a selective β2-adrenergic receptor antagonist, supplied for research use to study β2 receptor pharmacology and related physiological effects. It is provided as a high-purity solid suitable for preclinical and biochemical applications.
  • Selective β2-adrenergic receptor antagonist for receptor pharmacology studies.
  • Hydrochloride salt form to aid solubility and handling.
  • Reported high purity (~99.9%) for reliable experimental use.
  • Available in small research pack sizes for preclinical experimentation.
  • Identified by CAS 5696-15-1 and molecular weight 303.82 for unambiguous reference.
Catalog No. 50-002-27456 Supplier Medchemexpress LLC Supplier No. HY11847010MG
Butaxamine hydrochloride is the hydrochloride salt of a selective β2-adrenergic receptor antagonist, supplied for research use to study β2 receptor pharmacology and related physiological effects. It is provided as a high-purity solid suitable for preclinical and biochemical applications.
  • Selective β2-adrenergic receptor antagonist for receptor pharmacology studies.
  • Hydrochloride salt form to aid solubility and handling.
  • Reported high purity (~99.9%) for reliable experimental use.
  • Available in small research pack sizes for preclinical experimentation.
  • Identified by CAS 5696-15-1 and molecular weight 303.82 for unambiguous reference.